EMPIGEN BB ~30% (aktive Substanz), Sigma-Aldrich®
Lieferant: Merck
Synonyme: N,N-Dimethyl-N-dodecylglycine betaine, N-(Alkyl C10-C16)-N,N-dimethylglycine betaine
M-Clarity™ Qualitätsstufe = MQ200
30326-250ML 30326-50ML
SIAL30326-250MLEA 136 EUR
SIAL30326-250ML SIAL30326-50ML
EMPIGEN BB ~30% (aktive Substanz), Sigma-Aldrich®
EMPIGEN® BB detergent
Empigen BB is a zwitterionic detergent compatible with anionic, non-ionic, cationic and other amphoteric surfactants.
Empigen BB is a useful detergent that has been used in the solubilization of membrane proteins and biochemical analysis of keratins. It not only helps to solubilize bigger amounts of protein but it also maintains antibody reactivity. It has been also used in the purification of leucine-rich repeat kinase 2 (LRRK2) in Escherichia coli during research in Parkinson′s disease. The use of Empigen in the preparation of viral antigens is being increasing due to its effectivity in giving greater antibody responses.
Bitte beachten EMPIGEN is a registered trademark of Huntsman Corporation
Formel: CH₃(CH₂)₈-₁₄CH₂N+(CH₃)₂CH₂COO-
MDL Number: MFCD00084742
CAS-Nummer: 66455-29-6