QuEChERS standards
Lieferant: Agilent
5190-0501 5190-0499 5190-0503
HEWL5190-0501EA 382 EUR
HEWL5190-0501 HEWL5190-0499 HEWL5190-0503
QuEChERS standards
Standards Lebensmittel-Standards
Ready-to-use QuEChERS standards are provided for the most commonly used regulatory methods, including AOAC and EN making analysis easier on both GC and LC instruments.
  • Save time and inconvenience of making standards
  • Available for both GC and LC instruments
  • Ready to use for QuEChERS extractions – no dilutions required
In addition to our industry-leading QuEChERS Kits, Agilent makes your analysis easier by providing standards for the most commonly used regulatory methods, including AOAC and EN.