Sample cones for PerkinElmer
Lieferant: Agilent
8003-0571 8003-0573 8003-0575 8003-0577
HEWL8003-0571EA 649 EUR
HEWL8003-0571 HEWL8003-0573 HEWL8003-0575 HEWL8003-0577
Sample cones for PerkinElmer
Chromatographie-Zubehör Chromatographie-Ersatzteile
These sample and skimmer cones are equivalent to those from PerkinElmer ICP-MS for complete confidence.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
  • Many parts in Agilent packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage
Large-orifice sampler and skimmer cones provide superior long-term stability and resist clogging, allowing analysis under both high and low sample-uptake conditions. Nickel is a very rugged, long-lasting material.