Captiva EMR-Lipid cartridges
Lieferant: Agilent
5190-1003 5190-1004 5190-1002
HEWL5190-1003EA 404 EUR
HEWL5190-1003 HEWL5190-1004 HEWL5190-1002
Captiva EMR-Lipid cartridges
Chromatographiesäulen
Captiva EMR–Lipid provides highly selective and efficient lipid/matrix removal without unwanted analyte loss.
  • Highly selective and efficient lipid removal: The unique EMR Lipid mechanism combines size exclusion and hydrophobic interactions between the sorbent and the long aliphatic chain of lipids
  • Clog-free operation: Advanced filter design and construction technology ensures an easy flow
  • Time savings and improved precision: A protein crash solvent retention frit in 1 ml and 96-well plate formats promotes a streamlined, automatable in-well protein precipitation workflow
The novel EMR-Lipid technology removes lipids based on a combination of size exclusion and hydrophobic interaction. Effective lipid removal assures minimal ion suppression of target analytes, which significantly improves method reliability and ruggedness. The 96-well plate and 1 ml cartridge formats contain a solvent retention frit, which allows solvent-first in-well protein precipitation which streamlines sample preparation. The improved filter design gives easy flow with vacuum or positive pressure. The 3 and 6 ml cartridge formats allow gravity flow with the absence of solvent retention frits and are easy to use.