Vacuum supplies
Lieferant: Agilent
G1833-65332 5069-4478 9499342M002 9499392M004
HEWLG1833-65332EA 117 EUR
HEWLG1833-65332 HEWL5069-4478 HEWL9499342M002 HEWL9499392M004
Vacuum supplies
Chromatographie-Zubehör Chromatographie-Ersatzteile
A range of vacuum pump supplies is available to ensure the accuracy and performance of your ICP-MS vacuum system.
  • When plasma is ON, check pump oil level every three months
  • Maintain oil level between max and min levels on the gauge
  • Standard frequency for changing the oil is approximately every six months
Bitte beachten Be careful when changing oil because the oil may have absorbed dangerous substances. The oil in the rotary pump on the interface side of an ICP-MS instrument equipped with a hydrofluoric acid resisting introduction system is especially dangerous. Do not touch with bare hands.