Wasser, DEPC-behandelt
Lieferant: G Bioscienes
Synonyme: Hydrogen oxide
786-109 786-117 786-118
786-109EA 156 EUR
786-109 786-117 786-118
Wasser, DEPC-behandelt
Wasser
Ultra pure DEPC-Treated water is suitable for use with sensitive RNA research. It is prepared by incubating with 0.1% DEPC (diethylpyrocarbonate), an RNase inhibitor, and then autoclaved to remove the DEPC, to prevent downstream interference, and sterilized.
Formel: H₂O
Molekulargewicht: 18.02 g/mol
Siedepunkt: 100
Schmelzpunkt: 0
MDL Number: MFCD00011332
CAS-Nummer: 7732-18-5
Merck Index: 13,10098