Leupeptin hemisulphate
Lieferant: Apollo Scientific
BIMI2183-100MG
BIMI2183-25MG
BIMI2183-50MG
APOSBIMI2183-100MGEA
299
EUR
APOSBIMI2183-100MG
APOSBIMI2183-25MG
APOSBIMI2183-50MG
Leupeptin hemisulphate
Proteine und Peptide
Leupeptin is a peptide antibiotic, isolated from several Streptomyces species. It is a reversible inhibitor ofserine and cysteine proteases