Phosphoramidon disodium salt
Lieferant: Apollo Scientific
BIMI0549-10MG BIMI0549-5MG
APOSBIMI0549-10MGEA 284 EUR
APOSBIMI0549-10MG APOSBIMI0549-5MG
Phosphoramidon disodium salt
Proteine und Peptide
Inhibitor of thermolysin. Weak collagenase inhibitor. Does not inhibit chymotrypsin, papain, pepsin or trypsin.