2,6-Difluorbenzhydrazid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,6-Difluorobenzohydrazide
PC9953-1G
PC9953-10G
APOSPC9953-1GEA
58
EUR
APOSPC9953-1G
APOSPC9953-10G
2,6-Difluorbenzhydrazid
2,6-Difluorbenzhydrazid
Formel:
C₇H₆F₂N₂O Schmelzpunkt: 72…74 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00119406 CAS-Nummer: 172935-91-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...