5-Chloro-6-fluoro-1H-indole 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC9933-1G
PC9933-5G
APOSPC9933-1GEA
219
EUR
APOSPC9933-1G
APOSPC9933-5G
5-Chloro-6-fluoro-1H-indole 97%
5-Chloro-6-fluoro-1H-indole
Formel:
C8H5ClFN Schmelzpunkt: 83 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD01074495 CAS-Nummer: 169674-57-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...