2'-Fluor-6'-hydroxyacetophenon

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Fluoro-6-hydroxyphenyl)ethanone

PC9896-5G PC9896-1G PC9896-10G
APOSPC9896-5GEA 40.6 EUR
APOSPC9896-5G APOSPC9896-1G APOSPC9896-10G
2'-Fluor-6'-hydroxyacetophenon
2'-Fluor-6'-hydroxyacetophenon
Formel: C₈H₇FO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01090995
CAS-Nummer: 93339-98-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR