2-(4-Pyridinyl)-6-(trifluoromethyl)-4-pyrimidinol
Lieferant: Apollo Scientific
Synonyme:
4-Hydroxy-2-(pyridin-4-yl)-6-(trifluoromethyl)pyrimidine
PC9777-1G
PC9777-2.5G
APOSPC9777-1GEA
87.3
EUR
APOSPC9777-1G
APOSPC9777-2.5G
2-(4-Pyridinyl)-6-(trifluoromethyl)-4-pyrimidinol
2-(4-Pyridinyl)-6-(trifluoromethyl)-4-pyrimidinol
Formel:
C₁₀H₆F₃N₃O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00140054 CAS-Nummer: 204394-60-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...