[5-[(4-Fluorophenyl)sulphonyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol
Lieferant: Apollo Scientific
Synonyme:
5-(4-Fluorobenzenesulphonyl)-4-(hydroxymethyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole
PC9379-1G
PC9379-500MG
APOSPC9379-1GEA
666
EUR
APOSPC9379-1G
APOSPC9379-500MG
[5-[(4-Fluorophenyl)sulphonyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol
[5-[(4-Fluorophenyl)sulphonyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol
Formel:
C₁₂H₁₀F₄N₂O₃S Schmelzpunkt: 127…129 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00172654 CAS-Nummer: 318469-24-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...