3-(Difluormethoxy)benzaldehyd 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC9371-10G
PC9371-5G
PC9371-1G
APOSPC9371-10GEA
97.3
EUR
APOSPC9371-10G
APOSPC9371-5G
APOSPC9371-1G
3-(Difluormethoxy)benzaldehyd 99%
3-(Difluormethoxy)benzaldehyd
Formel:
C₈H₆F₂O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00236222 CAS-Nummer: 85684-61-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...