2-(3,4-Difluorphenyl)indol 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC9342-10G
PC9342-1G
PC9342-5G
APOSPC9342-10GEA
725
EUR
APOSPC9342-10G
APOSPC9342-1G
APOSPC9342-5G
2-(3,4-Difluorphenyl)indol 97%
2-(3,4-Difluorophenyl)indole
Formel:
C₁₄H₉F₂N Schmelzpunkt: 170…173 °C Lagerungstemperatur: Kühlschrank |
CAS-Nummer:
68290-36-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...