6-Fluor-o-toluylsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Fluoro-6-methylbenzoic acid
PC9286-1G
PC9286-25G
PC9286-5G
APOSPC9286-1GEA
21.9
EUR
APOSPC9286-1G
APOSPC9286-25G
APOSPC9286-5G
6-Fluor-o-toluylsäure 98%
6-Fluor-o-toluylsäure
Formel:
C₈H₇FO₂ Schmelzpunkt: 119…124 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06658162 CAS-Nummer: 90259-27-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...