2,3-Dibromo-5-fluoropyridine
Lieferant: Apollo Scientific
|
Achtung
|
PC9196-1G
PC9196-5G
APOSPC9196-1GEA
21.9
EUR
APOSPC9196-1G
APOSPC9196-5G
2,3-Dibromo-5-fluoropyridine
2,3-Dibromo-5-fluoropyridine
Formel:
C₅H₂Br₂FN Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD08532501 CAS-Nummer: 878207-82-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...