N-Fmoc-4-(trifluoromethyl)-L-phenylalanine

Lieferant: Apollo Scientific
Achtung

Synonyme: Fmoc-Phe(4-CF3)-OH

PC9134-1G PC9134-5G PC9134-250MG
APOSPC9134-1GEA 132 EUR
APOSPC9134-1G APOSPC9134-5G APOSPC9134-250MG
N-Fmoc-4-(trifluoromethyl)-L-phenylalanine
N-Fmoc-4-(trifluoromethyl)-L-phenylalanine
Formel: C₂₅H₂₀F₃NO₄
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD00797581
CAS-Nummer: 247113-86-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR