Boc-L-pentafluorophenylalanine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Boc-pentafluoro-Phe-OH, (S)-2-((tert-Butoxycarbonyl)amino)-3-(perfluorophenyl)propanoic acid
PC9113-1G
PC9113-25G
PC9113-5G
APOSPC9113-1GEA
102
EUR
APOSPC9113-1G
APOSPC9113-25G
APOSPC9113-5G
Boc-L-pentafluorophenylalanine
Boc-L-pentafluorophenylalanine
Formel:
C₁₄H₁₄F₅NO₄ Molekulargewicht: 355.261 g/mol Schmelzpunkt: 106…108 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00151893 CAS-Nummer: 34702-60-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...