[3-(4-Fluorophenoxy)propyl]amine 96%
Lieferant: Apollo Scientific
Synonyme:
3-(4-Fluorophenoxy)propylamine
PC911117-1G
PC911117-5G
APOSPC911117-1GEA
78.3
EUR
APOSPC911117-1G
APOSPC911117-5G
[3-(4-Fluorophenoxy)propyl]amine 96%
[3-(4-Fluorophenoxy)propyl]amine
Formel:
C₉H₁₂FNO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06213776 CAS-Nummer: 116753-54-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...