2-Brom-6-fluorimidazo[1,2-a]pyridin 97%

Lieferant: Apollo Scientific
Achtung

PC910708-100MG PC910708-250MG
APOSPC910708-100MGEA 71.2 EUR
APOSPC910708-100MG APOSPC910708-250MG
2-Brom-6-fluorimidazo[1,2-a]pyridin 97%
2-Brom-6-fluorimidazo[1,2-a]pyridin
Formel: C₇H₄BrFN₂
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD10698610
CAS-Nummer: 1060815-08-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR