3-Fluor-5-iodbenzoesäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Fluoro-3-iodobenzoic acid
PC9102-1G
PC9102-250MG
PC9102-5G
PC9102-25G
APOSPC9102-1GEA
21.9
EUR
APOSPC9102-1G
APOSPC9102-250MG
APOSPC9102-5G
APOSPC9102-25G
3-Fluor-5-iodbenzoesäure
3-Fluor-5-iodbenzoesäure
Formel:
C₇H₄FIO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD04116467 CAS-Nummer: 723294-74-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...