5-Fluoro-1,3-dimethyl-2-nitrobenzene 95%

Lieferant: Apollo Scientific

PC906351-1G PC906351-5G
APOSPC906351-1GEA 122 EUR
APOSPC906351-1G APOSPC906351-5G
5-Fluoro-1,3-dimethyl-2-nitrobenzene 95%
5-Fluoro-1,3-dimethyl-2-nitrobenzene
Formel: C₈H₈FNO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03413407
CAS-Nummer: 315-12-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR