4-Fluor-2,3-dimethylbenzaldehyd 98%

Lieferant: Apollo Scientific

Synonyme: 4-Fluoro-2,3-dimethylbenzaldehyde

PC901981-1G PC901981-5G
APOSPC901981-1GEA 290 EUR
APOSPC901981-1G APOSPC901981-5G
4-Fluor-2,3-dimethylbenzaldehyd 98%
4-Fluor-2,3-dimethylbenzaldehyd
Formel: C₉H₉FO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD16295095
CAS-Nummer: 363134-37-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR