1-(5-Fluoropyridin-2-yl)piperidine 98%

Lieferant: Apollo Scientific

Synonyme: 5-Fluoro-2-piperidinopyridine

PC900591-1G PC900591-5G
APOSPC900591-1GEA 145 EUR
APOSPC900591-1G APOSPC900591-5G
1-(5-Fluoropyridin-2-yl)piperidine 98%
1-(5-Fluoropyridin-2-yl)piperidine
Formel: C₁₀H₁₃FN₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD18089239
CAS-Nummer: 1287218-71-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR