3,5-Bis(trifluormethyl)benzamidin Hydrochlorid 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Bis(trifluoromethyl)benzene-1-carboximidamide hydrochloride, 3,5-Bis(trifluoromethyl)phenylamidine hydrochloride

PC9005-10G PC9005-1G
APOSPC9005-10GEA 473 EUR
APOSPC9005-10G APOSPC9005-1G
3,5-Bis(trifluormethyl)benzamidin Hydrochlorid 95+%
3,5-Bis(trifluormethyl)benzamidin Hydrochlorid
Formel: C₉H₇ClF₆N₂
Schmelzpunkt: 122…124 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00276604
CAS-Nummer: 97603-94-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR