4-Fluoro-2,6-dimethylbenzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,6-Dimethyl-4-fluorobenzoic acid

PC8961-1G PC8961-250MG PC8961-5G
APOSPC8961-1GEA 102 EUR
APOSPC8961-1G APOSPC8961-250MG APOSPC8961-5G
4-Fluoro-2,6-dimethylbenzoic acid
4-Fluoro-2,6-dimethylbenzoic acid
Formel: C₉H₉FO₂
Schmelzpunkt: 144…145 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06208279
CAS-Nummer: 16633-50-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR