4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carbohydrazide

Lieferant: Apollo Scientific

PC8924-1G PC8924-5G
APOSPC8924-1GEA 538 EUR
APOSPC8924-1G APOSPC8924-5G
4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carbohydrazide
4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carbohydrazide
Formel: C₁₂H₁₀F₃N₃OS
Schmelzpunkt: 170…173 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00112085
CAS-Nummer: 263387-25-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR