1,2-Difluoro-4-phenylsulphonylbenzene

Lieferant: Apollo Scientific

PC8856-1G PC8856-5G
APOSPC8856-1GEA 137 EUR
APOSPC8856-1G APOSPC8856-5G
1,2-Difluoro-4-phenylsulphonylbenzene
1,2-Difluoro-4-phenylsulphonylbenzene
Formel: C₁₂H₈F₂O₂S
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09835175
CAS-Nummer: 934241-80-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR