2-(4-Fluorophenyl)pyrrolidine 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC8814-1G
PC8814-250MG
APOSPC8814-1GEA
50.9
EUR
APOSPC8814-1G
APOSPC8814-250MG
2-(4-Fluorophenyl)pyrrolidine 97%
2-(4-Fluorophenyl)pyrrolidine
Formel:
C₁₀H₁₂FN Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD02663411 CAS-Nummer: 72216-06-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...