N-Ethyl-2-nitro-4-(trifluormethyl)anilin

Lieferant: Apollo Scientific
Achtung

PC8794-1G PC8794-250MG PC8794-5G
APOSPC8794-1GEA 79.9 EUR
APOSPC8794-1G APOSPC8794-250MG APOSPC8794-5G
N-Ethyl-2-nitro-4-(trifluormethyl)anilin
N-Ethyl-2-nitro-4-(trifluormethyl)anilin
Formel: C₉H₉F₃N₂O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00044178
CAS-Nummer: 30377-62-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR