3-Fluoro-5-iodo-4-methylbenzenecarboxylic acid 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Fluoro-5-iodo-4-methylbenzoic acid, 3-Fluoro-4-methyl-5-iodobenzoic acid
PC8697-1G
PC8697-500MG
APOSPC8697-1GEA
61.3
EUR
APOSPC8697-1G
APOSPC8697-500MG
3-Fluoro-5-iodo-4-methylbenzenecarboxylic acid 95+%
3-Fluoro-5-iodo-4-methylbenzenecarboxylic acid
Formel:
C₈H₆FIO₂ Schmelzpunkt: 171…173 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD16622791 CAS-Nummer: 861905-94-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...