2-(4-Fluorophenyl)-2-oxoethyl thiocyanate

Lieferant: Apollo Scientific
Achtung

PC8588-10G PC8588-1G PC8588-5G
APOSPC8588-10GEA 894 EUR
APOSPC8588-10G APOSPC8588-1G APOSPC8588-5G
2-(4-Fluorophenyl)-2-oxoethyl thiocyanate
2-(4-Fluorophenyl)-2-oxoethyl thiocyanate
Formel: C₉H₆FNOS
Schmelzpunkt: 99…101 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD02029655
CAS-Nummer: 43045-16-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR