5-Fluor-3-iod-1H-indazol 97%

Lieferant: Apollo Scientific
Achtung

PC8399-5G PC8399-1G
APOSPC8399-5GEA 91.5 EUR
APOSPC8399-5G APOSPC8399-1G
5-Fluor-3-iod-1H-indazol 97%
5-Fluoro-3-iodo-1H-indazole
Formel: C₇H₄FIN₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06739145
CAS-Nummer: 858629-06-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR