2,3,5-Trifluoro-4-pyridinamine 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC8305-1G
PC8305-250MG
APOSPC8305-1GEA
797
EUR
APOSPC8305-1G
APOSPC8305-250MG
2,3,5-Trifluoro-4-pyridinamine 97%
Formel:
C₅H₃F₃N₂ Schmelzpunkt: 103…105 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01862081 CAS-Nummer: 105252-95-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...