tert-Butyldiphenylsilyl trifluoromethanesulphonate

Lieferant: Apollo Scientific
Gefahr

Synonyme: tert-Butyldiphenylsilyl Triflate, Trifluoromethanesulphonic acid tert-Butyldiphenylsilyl Ester

PC7942-1G PC7942-5G
APOSPC7942-1GEA 30.6 EUR
APOSPC7942-1G APOSPC7942-5G
tert-Butyldiphenylsilyl trifluoromethanesulphonate
tert-Butyldiphenylsilyl trifluoromethanesulphonate
Formel: C₁₇H₁₉F₃O₃SSi
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD01941212
CAS-Nummer: 92886-86-7
UN-Nummer: 1760
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR