3-Brom-2,6-difluortoluol 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC7858-25G
PC7858-5G
APOSPC7858-25GEA
94.8
EUR
APOSPC7858-25G
APOSPC7858-5G
3-Brom-2,6-difluortoluol 99%
Formel:
C₇H₅BrF₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03412221 CAS-Nummer: 221220-97-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...