3,5-Dichlor-2,4-difluoranilin 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC7783-5G
PC7783-1G
PC7783-25G
PC7783-100G
APOSPC7783-5GEA
21.9
EUR
APOSPC7783-5G
APOSPC7783-1G
APOSPC7783-25G
APOSPC7783-100G
3,5-Dichlor-2,4-difluoranilin 97%
3,5-Dichlor-2,4-difluoranilin
Formel:
C₆H₃Cl₂F₂N Schmelzpunkt: 75 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00044104 CAS-Nummer: 83121-15-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...