(α,α,α-Trifluor-3-tolyl)essigsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-(Trifluoromethyl)phenyl)acetic acid, 3-(Trifluoromethyl)phenylacetic acid
PC7699-25G
PC7699-100G
PC7699-5G
APOSPC7699-25GEA
34.8
EUR
APOSPC7699-25G
APOSPC7699-100G
APOSPC7699-5G
(α,α,α-Trifluor-3-tolyl)essigsäure 97%
(α,α,α-Trifluor-3-tolyl)essigsäure
Formel:
(C₆H₄(CF₃))CH₂COOH Schmelzpunkt: 76…79 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00004339 CAS-Nummer: 351-35-9 EINECS: 206-511-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...