4-(Difluormethoxy)phenylessigsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-(Difluoromethoxy)phenyl)acetic acid

PC7690-1G PC7690-250MG PC7690-5G
APOSPC7690-1GEA 66.7 EUR
APOSPC7690-1G APOSPC7690-250MG APOSPC7690-5G
4-(Difluormethoxy)phenylessigsäure 98%
4-(Difluormethoxy)phenylessigsäure
Formel: C₉H₈F₂O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00236239
CAS-Nummer: 243659-15-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR