Benzyl chlorofluoroacetate 97%

Lieferant: Apollo Scientific

PC7636-1G PC7636-5G PC7636-25G
APOSPC7636-1GEA 281 EUR
APOSPC7636-1G APOSPC7636-5G APOSPC7636-25G
Benzyl chlorofluoroacetate 97%
Benzyl chlorofluoroacetate
Formel: C₉H₈ClFO₂
Siedepunkt: 125…126 °C/15mmHg
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01862039
CAS-Nummer: 243659-11-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR