2,4-Difluor-5-methoxyanilin 99%

Lieferant: Apollo Scientific
Achtung

PC7611-1G PC7611-500MG PC7611-5G
APOSPC7611-1GEA 346 EUR
APOSPC7611-1G APOSPC7611-500MG APOSPC7611-5G
2,4-Difluor-5-methoxyanilin 99%
2,4-Difluor-5-methoxyanilin
Formel: C₇H₇NOF₂
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD08689699
CAS-Nummer: 98446-51-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR