2-Amino-4-chlor-2'-fluorbenzophenon

Lieferant: Apollo Scientific
Achtung

PC7608-1G PC7608-250MG
APOSPC7608-1GEA 495 EUR
APOSPC7608-1G APOSPC7608-250MG
2-Amino-4-chlor-2'-fluorbenzophenon
2-Amino-4-chlor-2'-fluorbenzophenon
Formel: C₁₃H₉ClFNO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00236584
CAS-Nummer: 203303-05-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR