2-Brom-N-(2,4-difluorphenyl)benzolsulfonamid 98%
Lieferant: Apollo Scientific
PC7584-1G
PC7584-5G
APOSPC7584-1GEA
132
EUR
APOSPC7584-1G
APOSPC7584-5G
2-Brom-N-(2,4-difluorphenyl)benzolsulfonamid 98%
2-Brom-N-(2,4-difluorphenyl)benzolsulfonamid
Formel:
C₁₂H₈BrF₂NO₂S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09475878 CAS-Nummer: 941294-30-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...