3-Ethoxy-4-fluorophenylboronic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Ethoxy-4-fluorobenzeneboronic acid

PC7571-1G PC7571-5G PC7571-10G
APOSPC7571-1GEA 142 EUR
APOSPC7571-1G APOSPC7571-5G APOSPC7571-10G
3-Ethoxy-4-fluorophenylboronic acid 95%
3-Ethoxy-4-fluorophenylboronic acid
Formel: C₈H₁₀BFO₃
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD09475831
CAS-Nummer: 900174-65-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR