2-Brom-5-fluor-4-methylanilin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-5-fluoro-p-toluidine

PC7405-1G PC7405-25G PC7405-5G
APOSPC7405-1GEA 21.9 EUR
APOSPC7405-1G APOSPC7405-25G APOSPC7405-5G
2-Brom-5-fluor-4-methylanilin 98%
2-Brom-5-fluor-4-methylanilin
Formel: C₇H₇BrFN
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00142871
CAS-Nummer: 202865-78-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR