2-Fluor-5-nitro-6-picolin 98%
Lieferant: Apollo Scientific
PC7203-10G
PC7203-25G
APOSPC7203-10GEA
77.1
EUR
APOSPC7203-10G
APOSPC7203-25G
2-Fluor-5-nitro-6-picolin 98%
2-Fluor-5-nitro-6-picolin
Formel:
C₆H₅FN₂O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03095071 CAS-Nummer: 18605-16-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...