6-Brom-2,3-difluortoluol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-3,4-difluoro-2-methylbenzene
PC7027-1G
PC7027-5G
PC7027-25G
APOSPC7027-1GEA
27.7
EUR
APOSPC7027-1G
APOSPC7027-5G
APOSPC7027-25G
6-Brom-2,3-difluortoluol
6-Brom-2,3-difluortoluol
Formel:
C₇H₅BrF₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD07777161 CAS-Nummer: 847502-81-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...