Bis(pyridin)iodoniumtetrafluoroborat
Lieferant: Apollo Scientific
|
Achtung
|
PC6427-1G
PC6427-5G
APOSPC6427-1GEA
65.4
EUR
APOSPC6427-1G
APOSPC6427-5G
Bis(pyridin)iodoniumtetrafluoroborat
Formel:
C₁₀H₁₀BF₄IN₂ Schmelzpunkt: 159…164 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD03703393 CAS-Nummer: 15656-28-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...