4-Nitro-5-(trifluoromethyl)-2-[2-(trimethylsilyl)ethynyl]phenylamine
Lieferant: Apollo Scientific
Synonyme:
5-Amino-2-nitro-4-[2-(trimethylsilyl)ethynyl]benzotrifluoride
PC6111-1G
PC6111-500MG
APOSPC6111-1GEA
538
EUR
APOSPC6111-1G
APOSPC6111-500MG
4-Nitro-5-(trifluoromethyl)-2-[2-(trimethylsilyl)ethynyl]phenylamine
4-Nitro-5-(trifluoromethyl)-2-[2-(trimethylsilyl)ethynyl]phenylamine
Formel:
C₁₂H₁₃F₃N₂O₂Si Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD12827775 CAS-Nummer: 1186405-13-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...